1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate 98%

Warning

L00956-09 L00956-18 L00956-18 L00956-09
AAAL00956-09EA 41.05 USD
AAAL00956-09 CAAAAL00956-09 AAAL00956-18 CAAAAL00956-18
1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate 98%
1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate
Formula: C₃₈H₃₂Br₂P₂·1H₂O
MW: 728.45 g/mol (710.44 anhy)
Melting Pt: ca 236…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00040548
CAS Number: 54770-27-3
EINECS: 259-334-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR