Ethyl-2-butylacetoacetate 95%

Synonyms: Ethyl 2-acetylhexanoate2-Butylacetoacetic Acid Ethyl EsterEthyl 2-Acetylhexanoate2-Acetylhexanoic Acid Ethyl Ester

L00905-06 L00905-06
AAAL00905-06EA 61.39 USD
CAAAAL00905-06 AAAL00905-06
Ethyl-2-butylacetoacetate 95%
Ethyl-2-butylacetoacetate
Formula: C₁₀H₁₈O₃
MW: 186.25 g/mol
Boiling Pt: 224 °C
Density: 0.950
Storage Temperature: Ambient
MDL Number: MFCD00026888
CAS Number: 1540-29-0
EINECS: 216-271-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR