(Trimethylsilylmethyl)triphenylphosphonium iodide 98%

Warning

L00788-06 L00788-03 L00788-06 L00788-03
AAAL00788-03EA 38.38 USD
CAAAAL00788-03 AAAL00788-03 CAAAAL00788-06 AAAL00788-06
(Trimethylsilylmethyl)triphenylphosphonium iodide 98%
(Trimethylsilylmethyl)triphenylphosphonium iodide
Formula: C₂₂H₂₆IPSi
Melting Pt: ca 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051853
CAS Number: 3739-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR