(3,3-Dimethylallyl)triphenylphosphonium bromide 98+%

Warning

Synonyms: (3-Methyl-2-butenyl)triphenyl-phosphonium Bromide

L00513-06 L00513-14 L00513-06 L00513-14
AAAL00513-14EA 176.49 USD
CAAAAL00513-14 AAAL00513-14 AAAL00513-06 CAAAAL00513-06
(3,3-Dimethylallyl)triphenylphosphonium bromide 98+%
(3,3-Dimethylallyl)triphenylphosphonium bromide
Formula: C₂₃H₂₄BrP
MW: 411.33 g/mol
Melting Pt: 235…239 °C
Storage Temperature: Ambient
MDL Number: MFCD00051864
CAS Number: 1530-34-3
EINECS: 216-224-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR