4-Nitrophenyloxamic acid 98%

Warning

L00325-03 L00325-06 L00325-03 L00325-06
AAAL00325-03EA 45.13 USD
CAAAAL00325-06 AAAL00325-03 CAAAAL00325-03 AAAL00325-06
4-Nitrophenyloxamic acid 98%
4-Nitrophenyloxamic acid
Formula: C₈H₆N₂O₅
Melting Pt: 205 °C (dec.)
Storage Temperature: Ambient
EINECS: 203-160-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR