Phencyclone 97%

Supplier: Thermo Scientific Chemicals

L00108-06 L00108-03 L00108-06 L00108-03
AAAL00108-03EA 56.89 USD
CAAAAL00108-06 AAAL00108-03 CAAAAL00108-03 AAAL00108-06
Phencyclone 97%
Phencyclone
Formula: C₂₉H₁₈O
MW: 382.46 g/mol
Melting Pt: 245…249 °C
Storage Temperature: Ambient
MDL Number: MFCD00051451
CAS Number: 5660-91-3
EINECS: 227-112-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR