1-(3-Methylpyridin-2-yl)-1,4-diazepane 95%

Danger

H51006-03 H51006-03 H51006-MD H51006-MD
AAAH51006-MDEA 80.93 USD
AAAH51006-MD CAAAAH51006-MD AAAH51006-03 CAAAAH51006-03
1-(3-Methylpyridin-2-yl)-1,4-diazepane 95%
1-(3-Methylpyridin-2-yl)-1,4-diazepane
Formula: C₁₁H₁₇N₃
Storage Temperature: Ambient
MDL Number: MFCD08061111
CAS Number: 880362-05-0
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR