3-(2-Chloro-5-methylphenoxy)piperidine 98%

Warning

H50991-MD H50991-03 H50991-03 H50991-MD
AAAH50991-03EA 187.59 USD
CAAAAH50991-03 AAAH50991-03 CAAAAH50991-MD AAAH50991-MD
3-(2-Chloro-5-methylphenoxy)piperidine 98%
3-(2-Chloro-5-methylphenoxy)piperidine
Formula: C₁₂H₁₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD08061103
CAS Number: 902837-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR