2-(1-Piperazinyl)nicotinamide 95%

Warning

H50967-03 H50967-03 H50967-MD H50967-MD
AAAH50967-03EA 228.69 USD
CAAAAH50967-03 CAAAAH50967-MD AAAH50967-03 AAAH50967-MD
2-(1-Piperazinyl)nicotinamide 95%
2-(1-Piperazinyl)nicotinamide
Formula: C₁₀H₁₄N₄O
Storage Temperature: Ambient
MDL Number: MFCD08061093
CAS Number: 87394-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR