4-(4-Methylphenoxy)piperidine 99%
|
Warning
|
H50965-MD
H50965-MD
H50965-03
H50965-03
AAAH50965-03EA
317.21
USD
CAAAAH50965-03
AAAH50965-03
CAAAAH50965-MD
AAAH50965-MD
4-(4-Methylphenoxy)piperidine 99%
4-(4-Methylphenoxy)piperidine
Formula:
C₁₂H₁₇NO Storage Temperature: Ambient |
MDL Number:
MFCD04115017 CAS Number: 63843-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...