2-(4-Piperidinyloxy)benzonitrile 98%

Warning

H50961-MD H50961-MD H50961-03 H50961-03
AAAH50961-03EA 246.42 USD
AAAH50961-03 CAAAAH50961-03 AAAH50961-MD CAAAAH50961-MD
2-(4-Piperidinyloxy)benzonitrile 98%
2-(4-Piperidinyloxy)benzonitrile
Formula: C₁₂H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD08061090
CAS Number: 900572-37-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR