4-(3-Chlorophenoxy)piperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

H50956-MD H50956-03 H50956-MD H50956-03
AAAH50956-03EA 310.99 USD
CAAAAH50956-03 CAAAAH50956-MD AAAH50956-03 AAAH50956-MD
4-(3-Chlorophenoxy)piperidine 98%
4-(3-Chlorophenoxy)piperidine
Formula: C₁₁H₁₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD08061087
CAS Number: 97840-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR