4-(2-Methylphenoxy)piperidine 97%

Warning

H50951-MD H50951-03 H50951-MD H50951-03
AAAH50951-MDEA 81.14 USD
AAAH50951-MD CAAAAH50951-03 AAAH50951-03 CAAAAH50951-MD
4-(2-Methylphenoxy)piperidine 97%
4-(2-Methylphenoxy)piperidine
Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD06248344
CAS Number: 63843-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR