Ethyl-3-formylbenzoate 98%
|
Warning
|
H50945-03
H50945-MD
H50945-MD
H50945-03
AAAH50945-MDEA
95.46
USD
CAAAAH50945-03
AAAH50945-MD
AAAH50945-03
CAAAAH50945-MD
Ethyl-3-formylbenzoate 98%
Ethyl 3-formylbenzoate
Formula:
C₁₀H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD08064275 CAS Number: 33745-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...