3-(3-Piperidinyloxy)benzonitrile 99%

Supplier: Thermo Scientific Chemicals

Warning

H50939-03 H50939-03 H50939-MD H50939-MD
AAAH50939-03EA 231 USD
CAAAAH50939-03 AAAH50939-03 CAAAAH50939-MD AAAH50939-MD
3-(3-Piperidinyloxy)benzonitrile 99%
3-(3-Piperidinyloxy)benzonitrile
Formula: C₁₂H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD08061079
CAS Number: 902836-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR