2-Chloro-4-(3-piperidinyloxy)benzonitrile 97%

Warning

H50938-MD H50938-MD H50938-03 H50938-03
AAAH50938-03EA 267.4 USD
AAAH50938-03 CAAAAH50938-03 CAAAAH50938-MD AAAH50938-MD
2-Chloro-4-(3-piperidinyloxy)benzonitrile 97%
2-Chloro-4-(3-piperidinyloxy)benzonitrile
Formula: C₁₂H₁₃ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD08061078
CAS Number: 902836-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR