3-Phenoxypiperidine 97%

Supplier: Thermo Scientific Chemicals

Warning

H50912-03 H50912-03 H50912-MD H50912-MD
AAAH50912-03EA 285 USD
AAAH50912-03 CAAAAH50912-MD AAAH50912-MD CAAAAH50912-03
3-Phenoxypiperidine 97%
3-Phenoxypiperidine
Formula: C₁₁H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD04117746
CAS Number: 151666-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR