3-Benzyloxyphenylacetic acid 95%

Warning

Synonyms: 2-(3-(Benzyloxy)phenyl)acetic acid

H50908-03 H50908-MD H50908-MD H50908-03
AAAH50908-MDEA 66.15 USD
CAAAAH50908-03 AAAH50908-MD AAAH50908-03 CAAAAH50908-MD
3-Benzyloxyphenylacetic acid 95%
3-Benzyloxyphenylacetic acid
Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664806
CAS Number: 1860-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR