(2,6-Dimethoxyphenoxy)acetic acid ≥98%

Warning

H50894-MD H50894-03 H50894-MD H50894-06 H50894-03
AAAH50894-MDEA 28.39 USD
AAAH50894-MD CAAAAH50894-MD AAH50894-06 AAAH50894-03 CAAAAH50894-03
(2,6-Dimethoxyphenoxy)acetic acid ≥98%
(2,6-Dimethoxyphenoxy)acetic acid
Formula: C₁₀H₁₂O₅
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD06823823
CAS Number: 95110-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR