3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine 97%

Warning

H50879-MD H50879-MD H50879-03 H50879-03
AAAH50879-03EA 268.57 USD
AAAH50879-03 CAAAAH50879-MD AAAH50879-MD CAAAAH50879-03
3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine 97%
3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine
Formula: C₉H₁₅N₃O
MW: 181.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061044
CAS Number: 139269-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR