3-Methyl-2-(1-piperazinyl)quinoline 97%

Warning

H50834-MD H50834-MD H50834-03 H50834-03
AAAH50834-03EA 299.07 USD
CAAAAH50834-03 AAAH50834-03 CAAAAH50834-MD AAAH50834-MD
3-Methyl-2-(1-piperazinyl)quinoline 97%
3-Methyl-2-(1-piperazinyl)quinoline
Formula: C₁₄H₁₇N₃
MW: 227.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061017
CAS Number: 348133-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR