4-[4-(Trifluoromethoxy)phenoxy]piperidine 97%

Warning

Synonyms: 4-(4-Trifluoromethoxyphenoxy)piperidine

H50812-03 H50812-MD H50812-MD H50812-03
AAAH50812-MDEA 82.32 USD
CAAAAH50812-03 AAAH50812-MD AAAH50812-03 CAAAAH50812-MD
4-[4-(Trifluoromethoxy)phenoxy]piperidine 97%
4-[4-(Trifluoromethoxy)phenoxy]piperidine
Formula: C₁₂H₁₄F₃NO₂
MW: 261.25 g/mol
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD06656166
CAS Number: 287952-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR