2-(Isopropylamino)acetophenone hydrochloride 98%

Warning

H50799-03 H50799-MD H50799-MD H50799-03
AAAH50799-03EA 187.59 USD
AAAH50799-03 CAAAAH50799-03 AAAH50799-MD CAAAAH50799-MD
2-(Isopropylamino)acetophenone hydrochloride 98%
2-(Isopropylamino)acetophenone hydrochloride
Formula: C₁₁H₁₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD18785640
CAS Number: 38506-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR