4-(2-Chloro-5-methylphenoxy)piperidine 99%

Warning

H50778-MD H50778-MD H50778-03 H50778-03
AAAH50778-MDEA 72.96 USD
AAAH50778-MD CAAAAH50778-MD AAAH50778-03 CAAAAH50778-03
4-(2-Chloro-5-methylphenoxy)piperidine 99%
4-(2-Chloro-5-methylphenoxy)piperidine
Formula: C₁₂H₁₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD06246346
CAS Number: 254883-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR