N-Methyl-2-biphenylmethylamine 99%

Warning

H50765-03 H50765-03 H50765-MD H50765-MD
AAAH50765-MDEA 86.04 USD
AAAH50765-MD CAAAAH50765-MD AAAH50765-03 CAAAAH50765-03
N-Methyl-2-biphenylmethylamine 99%
N-Methyl-2-biphenylmethylamine
Formula: C₁₄H₁₅N
MW: 197.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00598943
CAS Number: 13737-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR