1-(4-Methoxyphenyl)-2-methylaminoethanol 97%

Warning

H50754-03 H50754-03 H50754-MD
AAAH50754-03EA 255.66 USD
CAAAAH50754-03 AAAH50754-03 CAAAAH50754-MD
1-(4-Methoxyphenyl)-2-methylaminoethanol 97%
1-(4-Methoxyphenyl)-2-methylaminoethanol
Formula: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD08060969
CAS Number: 58777-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR