3-(1-Piperazino)propionitrile

Warning

H50710-06 H50710-03 H50710-06 H50710-03
AAAH50710-06EA 261.53 USD
AAAH50710-06 AAAH50710-03 CAAAAH50710-06 CAAAAH50710-03
3-(1-Piperazino)propionitrile
3-(1-Piperazino)propionitrile
Formula: C₇H₁₃N₃
MW: 139.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093566
CAS Number: 34064-86-3
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR