3-(4-Chlorostyryl)-1H-pyrazole

Supplier: Thermo Scientific Chemicals
Warning

H50660-06 H50660-03 H50660-03 H50660-06
AAAH50660-06EA 392.94 USD
CAAAAH50660-03 CAAAAH50660-06 AAAH50660-06 AAAH50660-03
3-(4-Chlorostyryl)-1H-pyrazole
3-(4-Chlorostyryl)-1H-pyrazole
Formula: C₁₁H₉ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00120939
CAS Number: 852691-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR