3-(2,4-Dimethoxyphenyl)-1H-pyrazole 98%

Warning

H50637-03 H50637-06 H50637-06 H50637-03
AAAH50637-03EA 171.23 USD
CAAAAH50637-06 AAAH50637-03 AAAH50637-06 CAAAAH50637-03
3-(2,4-Dimethoxyphenyl)-1H-pyrazole 98%
3-(2,4-Dimethoxyphenyl)-1H-pyrazole
Formula: C₁₁H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02091529
CAS Number: 150433-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR