5-Bromo-2-isopropoxybenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H50627-03 H50627-MD H50627-MD H50627-03
AAAH50627-MDEA 55.14 USD
AAAH50627-MD CAAAAH50627-MD AAAH50627-03 CAAAAH50627-03
5-Bromo-2-isopropoxybenzonitrile 97%
5-Bromo-2-isopropoxybenzonitrile
Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02257441
CAS Number: 515832-52-7
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR