5-Bromo-2-ethoxybenzonitrile 99%

Warning

H50624-03 H50624-MD H50624-03 H50624-MD
AAAH50624-03EA 260.85 USD
CAAAAH50624-03 AAAH50624-03 CAAAAH50624-MD AAAH50624-MD
5-Bromo-2-ethoxybenzonitrile 99%
5-Bromo-2-ethoxybenzonitrile
Formula: C₈H₆BrNO
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD02257397
CAS Number: 279263-03-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR