4-Bromo-4'-methylbiphenyl 97%

Synonyms: 4-Bromo-4'-methyl-1,1'-biphenyl

H50596-03 H50596-MD H50596-03 H50596-MD
AAAH50596-03EA 394.05 USD
CAAAAH50596-03 AAAH50596-03 CAAAAH50596-MD AAAH50596-MD
4-Bromo-4'-methylbiphenyl 97%
4-Bromo-4'-methylbiphenyl
Formula: C₁₃H₁₁Br
MW: 247.14 g/mol
Melting Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00466236
CAS Number: 50670-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR