2-Bromo-N-methylbenzamide 99%

Warning

H50583-03 H50583-06 H50583-03 H50583-06
AAAH50583-03EA 138.4 USD
CAAAAH50583-03 AAAH50583-03 CAAAAH50583-06 AAAH50583-06
2-Bromo-N-methylbenzamide 99%
2-Bromo-N-methylbenzamide
Formula: C₈H₈BrNO
MW: 214.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00099250
CAS Number: 61436-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR