5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide

Warning

H50546-03 H50546-MD H50546-MD H50546-03
AAAH50546-03EA 287.49 USD
CAAAAH50546-03 AAAH50546-03 AAAH50546-MD CAAAAH50546-MD
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
Formula: C₅H₈ClN₃O₂S
MW: 209.66
Storage Temperature: Ambient
MDL Number: MFCD00105874
CAS Number: 88398-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR