(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone 97%

Warning

Synonyms: 4-(2-Chloropyridine-3-carbonyl)morpholine

H50522-MD H50522-03 H50522-MD H50522-03
AAAH50522-MDEA 67.56 USD
AAAH50522-MD CAAAAH50522-03 CAAAAH50522-MD AAAH50522-03
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone 97%
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone
Formula: C₁₀H₁₁ClN₂O₂
MW: 226.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00454210
CAS Number: 53062-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR