2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine 98%
|
Warning
|
H50520-MD
H50520-03
H50520-03
H50520-MD
AAAH50520-MDEA
74.94
USD
AAAH50520-MD
CAAAAH50520-MD
AAAH50520-03
CAAAAH50520-03
2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine 98%
2-Chloro-3-cyano-6-(4-methoxyphenyl)pyridine
Formula:
C₁₃H₉ClN₂O MW: 244.68 g/mol Melting Pt: 148 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00215274 CAS Number: 306977-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...