Ethyl-4-chloro-6-methoxyquinoline-3-carboxylate 97%

Synonyms: 4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl esterEthyl 4-chloro-6-methoxyquinoline-3-carboxylate

H50515-03 H50515-MD H50515-03 H50515-MD
AAAH50515-MDEA 75.95 USD
CAAAAH50515-03 AAAH50515-MD CAAAAH50515-MD AAAH50515-03
Ethyl-4-chloro-6-methoxyquinoline-3-carboxylate 97%
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
Formula: C₁₃H₁₂ClNO₃
MW: 265.70 g/mol
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00173417
CAS Number: 22931-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR