Ethyl-4-chloro-8-methylquinoline-3-carboxylate

Synonyms: 4-Chloro-8-methyl-quinoline-3-carboxylic acid ethyl ester

H50510-MD H50510-03 H50510-03 H50510-MD
AAAH50510-MDEA 54.54 USD
CAAAAH50510-MD AAAH50510-MD CAAAAH50510-03 AAAH50510-03
Ethyl-4-chloro-8-methylquinoline-3-carboxylate
Ethyl 4-chloro-8-methylquinoline-3-carboxylate
Formula: C₁₃H₁₂ClNO₂
MW: 249.70
Storage Temperature: Ambient
MDL Number: MFCD00173399
CAS Number: 37041-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR