Ethyl-4-chloro-8-methoxyquinoline-3-carboxylate
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
H50509-MD
H50509-03
H50509-MD
H50509-03
AAAH50509-MDEA
74.52
USD
AAAH50509-MD
CAAAAH50509-03
CAAAAH50509-MD
AAAH50509-03
Ethyl-4-chloro-8-methoxyquinoline-3-carboxylate
Ethyl 4-chloro-8-methoxyquinoline-3-carboxylate
Formula:
C₁₃H₁₂ClNO₃ MW: 265.70 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00173395 CAS Number: 27568-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...