Ethyl-4-chloro-8-fluoroquinoline-3-carboxylate 97%
Synonyms:
Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate
H50507-MD
H50507-03
H50507-MD
H50507-03
AAAH50507-MDEA
90.3
USD
CAAAAH50507-MD
AAAH50507-MD
CAAAAH50507-03
AAAH50507-03
Ethyl-4-chloro-8-fluoroquinoline-3-carboxylate 97%
Ethyl 4-chloro-8-fluoroquinoline-3-carboxylate
Formula:
C₁₂H₉ClFNO₂ MW: 253.66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00173364 CAS Number: 56824-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...