3,5-Dichloro-4-hydroxybenzonitrile 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloroxynil

H50432-06 H50432-06 H50432-03 H50432-03
AAAH50432-03EA 152.99 USD
CAAAAH50432-03 AAAH50432-03 CAAAAH50432-06 AAAH50432-06
3,5-Dichloro-4-hydroxybenzonitrile 97+%
3,5-Dichloro-4-hydroxybenzonitrile
Formula: C₇H₃Cl₂NO
Melting Pt: 143…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00002177
CAS Number: 1891-95-8
EINECS: 217-572-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR