2-Phthalimidoethanesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)ethanesulphonylchloride

H50382-03 H50382-03 H50382-06 H50382-06
AAAH50382-06EA 356.53 USD
AAAH50382-06 CAAAAH50382-06 AAAH50382-03 CAAAAH50382-03
2-Phthalimidoethanesulfonyl chloride 97%
2-Phthalimidoethanesulfonyl chloride
Formula: C₁₀H₈ClNO₄S
MW: 273.7 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD01861217
CAS Number: 4403-36-5
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR