3-(Difluoromethoxy)benzylamine 98%

Danger

Synonyms: (3-(Difluoromethoxy)phenyl)methanamine

H50322-03 H50322-06 H50322-06 H50322-03
AAAH50322-03EA 168.8 USD
AAAH50322-03 CAAAAH50322-03 CAAAAH50322-06 AAAH50322-06
3-(Difluoromethoxy)benzylamine 98%
3-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236230
CAS Number: 244022-71-7
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR