5-(1-Pyrrolidinyl)-2-furaldehyde 98%

Warning

Synonyms: 5-(Pyrrolidin-1-yl)furan-2-carbaldehyde

H50316-03 H50316-MD H50316-MD H50316-03
AAAH50316-MDEA 102.97 USD
CAAAAH50316-MD AAAH50316-MD CAAAAH50316-03 AAAH50316-03
5-(1-Pyrrolidinyl)-2-furaldehyde 98%
5-(1-Pyrrolidinyl)-2-furaldehyde
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD02629619
CAS Number: 84966-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR