Ethyl-2-(2-formylphenoxy)acetate 99%

H50286-06 H50286-MD H50286-03 H50286-MD H50286-03
AAH50286-06EA 200.55 USD
AAH50286-06 AAAH50286-03 CAAAAH50286-03 AAAH50286-MD CAAAAH50286-MD
Ethyl-2-(2-formylphenoxy)acetate 99%
Ethyl-2-(2-formylphenoxy)acetate
Formula: C₁₁H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00052131
CAS Number: 41873-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR