N-Isopropylpropylamine 96%

Supplier: Thermo Scientific Chemicals

Danger

H50256-03 H50256-03
AAAH50256-03EA 311 USD
CAAAAH50256-03 AAAH50256-03
N-Isopropylpropylamine 96%
N-Isopropylpropylamine
Formula: C₆H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD00027095
EINECS: 244-688-2
UN: 2733
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR