2-Chloro-4-iodonicotinic acid 98%

Warning

H50049-06 H50049-03 H50049-03 H50049-06
AAAH50049-03EA 150.1 USD
CAAAAH50049-03 CAAAAH50049-06 AAAH50049-03 AAAH50049-06
2-Chloro-4-iodonicotinic acid 98%
2-Chloro-4-iodonicotinic acid
Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
Melting Pt: 162…167 °C
Storage Temperature: Ambient
MDL Number: MFCD05663520
CAS Number: 544671-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR