5-Bromo-1H-pyrrolo[2,3-b]pyridine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Bromo-7-azaindole

H50040-MD H50040-MD H50040-03 H50040-03
AAAH50040-03EA 56.29 USD
AAAH50040-03 CAAAAH50040-03 AAAH50040-MD CAAAAH50040-MD
5-Bromo-1H-pyrrolo[2,3-b]pyridine 96%
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 178…179 °C
Storage Temperature: Ambient
MDL Number: MFCD06659677
CAS Number: 183208-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR