2-[(4-Chlorophenyl)sulfonyl]acetic acid 98%

Warning

H27414-MC
AAH27414-MCEA 101.56 USD
AAH27414-MC
2-[(4-Chlorophenyl)sulfonyl]acetic acid 98%
2-[(4-Chlorophenyl)sulfonyl]acetic acid
Formula: C₈H₇ClO₄S
Melting Pt: ca 140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00201948
CAS Number: 3405-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR