6-Acetyl-2-Benzoxazolinone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Acetylbenzo[d]oxazol-2(3H)-one

H27368-06
AAH27368-06EA 168.99 USD
AAH27368-06
6-Acetyl-2-Benzoxazolinone 97%
6-Acetyl-2-Benzoxazolinone
Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 232…239 °C
Storage Temperature: Ambient
MDL Number: MFCD01664312
CAS Number: 54903-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR